Learn More
Aat BioquestSupplier Diversity Partner Tide Quencher™ 4WS azide [TQ4WS azide] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2068
TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®. TQ4WS has (a). much stronger absorption; (b). much higher quenching efficiency; and (c).
Catalog No. 50-195-3061