Learn More
Aat BioquestSupplier Diversity Partner Tide Quencher™ 2 CPG [TQ2 CPG] *1000 Å* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2204
TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G. TQ2 has (a). much stronger absorption; (b). much higher quenching efficiency; and (c).
Catalog No. 50-195-3095