Learn More
Aat BioquestSupplier Diversity Partner Tide Quencher™ 5 CPG [TQ5 CPG] *1000 Å* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2078
TQ5 is designed to be a superior quencher to Cy5, TF5 and Cy5.5. TQ5 has (a). much stronger absorption; (b). much higher quenching efficiency; and (c). versatile reactive forms with desired solubility for labeling oligonucleotides and peptides.
Catalog No. 50-195-3066