Learn More
Tie-1 Mouse anti-Human, Alexa Fluor 405, Clone: 8D12B10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237270AF405
Catalog No. NBP237270N
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Tie-1
8D12B10
Western Blot, ELISA, Immunohistochemistry
Purified
TIE1
Purified recombinant fragment of human TIE1 (AA: 385-607) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Cancer, Protein Kinase
Primary
7075
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human