Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-1 Mouse anti-Human, Clone: Cl. 6F12, Novus Biologicals™
Mouse Monoclonal Antibody
$212.24 - $493.00
Specifications
Antigen | Tie-1 |
---|---|
Clone | Cl. 6F12 |
Concentration | LYOPH |
Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry, ELISA 1 - 15 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
Tie-1 Monoclonal specifically detects Tie-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Tie-1 | |
LYOPH | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Protein Kinase | |
P35590 | |
7075 | |
recombinant human soluble TIE-1 protein | |
Primary | |
Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml. | |
The monoclonal antibody will detect native human TIE-1 in ELISA experiments and on the surface of different human cell types. |
Cl. 6F12 | |
Western Blot 1 - 2 ug/ml, Flow Cytometry, ELISA 1 - 15 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
TIE1 | |
IgG1 | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title