Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie-1 Mouse anti-Human, FITC, Clone: 8D12B10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237270F

Catalog No. NBP237270F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Tie-1
8D12B10
ELISA, Immunohistochemistry
Purified
TIE1
Purified recombinant fragment of human TIE1 (AA: 385-607) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Cancer, Protein Kinase
Primary
7075
ELISA, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit