Learn More
Tie-2 Mouse anti-Human, HRP, Clone: MM0018-21G7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11060986H
Catalog No. NB11060986H
Add to cart
Description
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Tie-2
MM0018-21G7
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
TEK
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified human extracellular domain of Tie2 fragment).
Protein G purified
Angiogenesis, Cancer, Phospho Specific, Protein Kinase
Primary
7010
Human
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
HRP
PBS with No Preservative
angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes human Tie2, no cross-reactivity with human Tie1.