Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIE1 Mouse anti-Human, Clone: 8D12D2, Abnova™
Mouse monoclonal antibody raised against partial recombinant human TIE1.
Supplier: Abnova Corporation MAB12262
Specifications
TIE1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
JTK14/TIE | |
Mouse | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
8D12D2 | |
Mouse monoclonal antibody raised against partial recombinant human TIE1. | |
In PBS (0.05% sodium azide) | |
TIE1 | |
Recombinant protein corresponding to amino acids 385-607 of human TIE1. | |
RUO | |
7075 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction