Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIE2 Chicken anti-Mouse, Rat, Polyclonal, FabGennix
Chicken Polyclonal Antibody
Supplier: Fabgennix Inc TIE2201Y
Description
TIE2 Polyclonal antibody specifically detects TIE2 in Mouse, Rat samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
TIE2 | |
Polyclonal | |
Unconjugated | |
TEK | |
AA517024; Angiopoietin-1 receptor; CD202B; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; Tie2; tie-2; tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1 | |
Synthetic peptide taken within amino acid region 540-590 on human Tyrosine-protein kinase receptor Tie-1 isoform 2 protein. | |
200 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgY |
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q02858 | |
Chicken | |
Affinity chromatography | |
RUO | |
21687, 89804 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction