Learn More
TIF-IA Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303065A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Required for efficient transcription initiation by RNA polymerase I. Required for the formation of the competent preinitiation complex (PIC). Dissociates from pol I as a consequence of transcription. In vitro, cannot activate transcription in a subsequent transcription reaction.Specifications
TIF-IA | |
Polyclonal | |
Unconjugated | |
RRN3 | |
A-270G1.2, TIFIA, RNA polymerase I-specific transcription initiation factor RRN3, TIF-IA, transcription initiation factor IA, transcription initiation factor TIF-IA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54700 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9NYV6 | |
RRN3 | |
Between 601 and 651 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.