Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGAR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA585436
Description
Keep as concentrated solution. Positive Control: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes a member of the kelch family of proteins, which are characterized by kelch repeat motifs and a POZ/BTB protein-binding domain. It is thought that kelch repeats are actin binding domains. However, the specific function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
TIGAR | |
Polyclonal | |
Unconjugated | |
TIGAR | |
TIGAR antibody; C12orf5 antibody; TP53-induced glycolysis and apoptosis regulator antibody; probable fructose-2;6-bisphosphatase TIGAR antibody; transactivated by NS3TP2 protein antibody; chromosome 12 open reading frame 5 antibody | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57103 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.71 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NQ88 | |
TIGAR | |
Recombit protein encompassing a sequence within the center region of human TIGAR. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title