Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGD5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30964025UL

 View more versions of this product

Catalog No. NB124182


Add to cart

Description

Description

TIGD5 Polyclonal antibody specifically detects TIGD5 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

TIGD5
Unconjugated
PBS buffer, 2% sucrose
tigger transposable element derived 5, tigger transposable element-derived protein 5
The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse TIGD5 (NP_848761). Peptide sequence PGSTARPPPPPAPGPRPRVAVKMTFRKAYSIKDKLQAIERVKGGERQASV
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
84948
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit