Learn More
Chums™ Tiger Glove Clip
Secure gloves and other forms of PPE or accessories to tool belts, vehicles, or workstations. Clips are constructed from durable thermoplastic for high abrasion resistance, heat resistance, and dielectric properties.
Manufacturer:  Chums™ 30058100
 View more versions of this product
Catalog No. 19-038-730
Add to cart
This item is not returnable. View return policy
Description
Description
  • Thermoplastic glove clips secure gloves and other forms of PPE to tool belts, vehicles, or workstations
  • Spring loaded jaws open 0.75 in. to allow for 2 in. of total depth
  • Tested to 3,000 continuous cycles without loss of function or weight rating
  • Cold temperature tested down to 5°F
  • Effectively holds up to 5 lb.
Specifications
Specifications
Glove Clip
Hanging/Storing PPE
Thermoplastic
No
Black
Ear Muffs, Gloves, Hard Hats, Knee Pads, Noise Indicators, and Small Respirators
0°F