Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chums™ Tiger Glove Clip
SDP

Catalog Number 19038731
Click to view available options
Color:
Lime Green
This item is not returnable. View return policy
This item is not returnable. View return policy

Secure gloves and other forms of PPE or accessories to tool belts, vehicles, or workstations. Clips are constructed from durable thermoplastic for high abrasion resistance, heat resistance, and dielectric properties.

  • Thermoplastic glove clips secure gloves and other forms of PPE to tool belts, vehicles, or workstations
  • Spring loaded jaws open 0.75 in. to allow for 2 in. of total depth
  • Tested to 3,000 continuous cycles without loss of function or weight rating
  • Cold temperature tested down to 5°F
  • Effectively holds up to 5 lb.
Color Lime Green
Disposable Reusable
Material Thermoplastic
Product Type Glove Clip
For Use With (Application) Hanging/Storing PPE
For Use With (Equipment) Ear Muffs, Gloves, Hard Hats, Knee Pads, Noise Indicators, and Small Respirators
Temperature (English) Min 0°F
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit