Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT, Armenian Hamster anti-Mouse, Clone: 4D4, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Specifications

Antigen TIGIT
Clone 4D4
Applications Flow Cytometry, Blocking Assay
Host Species Armenian Hamster
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016449
View Documents
Novus Biologicals
NBP2811160.025MG
0.025 mg N/A
NB016450
View Documents
Novus Biologicals
NBP2811160.2MG
0.2 mg N/A
Description

Description

TIGIT Monoclonal antibody specifically detects TIGIT in Mouse Samples. It is validated for Flow Cytometry, Blocking Assay.
Specifications

Specifications

TIGIT
Flow Cytometry, Blocking Assay
Mouse
Primary
4D4
Armenian Hamster
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit