Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT, Mouse anti-Human, Alexa Fluor 532, Clone: TIGIT/3017, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279927AF532

Catalog No. NB012737


Add to cart

Description

Description

TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TIGIT
TIGIT/3017
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TIGIT
Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
Human
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50mM Sodium Borate with 0.05% Sodium Azide
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Mouse
IgG2c Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit