Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Mouse anti-Human, Alexa Fluor 647, Clone: 741182, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB7898R100UG

Catalog No. FAB7898R100


Add to cart
This item is not returnable. View return policy

Description

Description

TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TIGIT
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q495A1
Mouse
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human TIGIT in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse TIGIT or recombinant human DNAM-1/CD226 is observed.
Flow Cytometry
741182
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
TIGIT
Mouse myeloma cell line NS0-derived recombinant human TIGIT Met1-Pro141 Accession # Q495A1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
201633
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)