Learn More
TIGIT Mouse anti-Human, Brilliant Violet 711, Clone: 741182, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 747839
Catalog No. BDB747839
Add to cart
Description
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TIGIT
741182
Brilliant Violet 711
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2b, κ
RUO
Primary
TIGIT