Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT, Mouse anti-Human, Clone: TIGIT/3106, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279794100UG
Description
TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
TIGIT | |
Monoclonal | |
0.2 mg/mL | |
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
Mouse | |
Protein A or G purified | |
RUO | |
201633 | |
Store at 4°C. | |
IgG2b κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
TIGIT/3106 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
TIGIT | |
Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) | |
100 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction