Learn More
Thermo Scientific™ Accessories for the Barnstead™ TII Type 2 Water System Available on GSA/VA Contract for Federal Government customers only.
Integrate the accessories for the Thermo Scientific™ Barnstear™ TII Type 2 Water System with the system itself for a more complete laboratory solution.
Manufacturer:  Thermo Scientific™ AY1406X2
 View more versions of this product
Catalog No. 09-036-21
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Integrate the accessories for the Thermo Scientific™ Barnstear™ TII Type 2 Water System such as distribution pumps and UV lamps with the system itself to create a more complete lab solution.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Barnstead™ TII Type 2 Water System
0.067 to 0.1 µS/cm
1 to 5ppb
No
19.5 in.
13.5 in.
100/240VAC
4° to 40°C
47cm
Smart Distribution pump
12L/hr.
100/240VAC 50/60Hz
15MΩ·cm at 25°C
tap
<1CFU/mL
30 to 100psig (2 to 6.9 bar)
49.5cm
34.3cm
50/60Hz
18.5 in.
Smart Distribution Pump
1L/min.
Distribution Pump