Learn More
Tocris Bioscience™ Tilfrinib Available on GSA/VA Contract for Federal Government customers only.
Potent and selective breast tumor kinase (Brk) inhibitor
$190.10 - $799.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
55-791-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5579/10
Each for $190.10
Add to cart
55-795-0 View Documents Tocris Bioscience™
5579/50
Each for $799.00
Add to cart
Description
Description
Potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.