Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R3 Safety™ Tillman™ Premium Elkskin Stick Welders Gloves Non-distribution product as customer accommodation.

Gloves are constructed of premium elkskin leather that stays soft even when it gets hot. The back is lined with cotton/foam for additional heat protection, while the palm is unlined for enhanced dexterity.

$41.89

Specifications

Color White
Glove Cuff Style Gauntlet
Grade Premium
Material Elkskin Leather
Product Type Specs Welding Gloves
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-819-962 Non-distribution product as customer accommodation.
View Documents
R3 Safety™
750L
Large
Pair for $41.89
Add to cart
 
Description

Description

  • Pearl premium elkskin stays soft even when hot
  • Durable stitching with DuPont™ Aramid Fibers for additional strength
  • Welted fingers protect stitching
  • Cotton/foam lined back for added heat protection
  • Unlined palm for enhanced dexterity
  • Extended, 14 in. gauntlet cuff
  • Reinforced thumb
Specifications

Specifications

White
Premium
Welding Gloves
Gauntlet
Elkskin Leather
750
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit