Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R3 Safety™ Tillman™ Truefit™ Goatskin Gloves Non-distribution product as customer accommodation.

$26.59

Specifications

Back Material Spandex™
Color Black, White, Gray
Material Leather
Finger Style Smooth
For Use With (Application) Ideal for warehouse handling, maintenance work, power tool handling
View More Specs

Products 2
Catalog Number Mfr. No. Circumference (English) Hand Circumference (Metric) Hand Material Price Quantity & Availability  
Catalog Number Mfr. No. Circumference (English) Hand Circumference (Metric) Hand Material Price Quantity & Availability  
19-819-959 Non-distribution product as customer accommodation.
View Documents
R3 Safety™
1470L
9 to 10 in. 22.8 to 25.4cm Leather
Pair for $26.59
Add to cart
 
19-819-960 Non-distribution product as customer accommodation.
View Documents
R3 Safety™
1470XL
10 to 11 in. 25.4 to 27.9cm Leather
Pair for $26.59
Add to cart
 
Description

Description

Specifications

Specifications

Spandex™
Leather
Ideal for warehouse handling, maintenance work, power tool handling
Reinforced thumb, Leather side bolsters
Reinforced
Black, White, Gray
Smooth
Hook and loop elastic
Rough-side-out double
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit