Learn More
TIM-1/KIM-1/HAVCR Mouse anti-Human, Alexa Fluor™ 405, Clone: 990605, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB17503V100UG
Catalog No. FB17503V100
Add to cart
This item is not returnable. View return policy
Description
Description
TIM-1/KIM-1/HAVCR Monoclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TIM-1/KIM-1/HAVCR
990605
Flow Cytometry 0.25-1 ug/10^6 cells
Q96D42
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Human embryonic kidney cell, HEK293-derived human TIM-1/KIM-1/HAVCR, Met1-Thr288, Accession No. Q96D42
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human TIM-1/KIM-1/HAVCR in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
26762.0
Human