Learn More
TIM-2 Rat anti-Mouse, PE, Clone: RMT2-26, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566591
Catalog No. BDB566591
Add to cart
Description
Description
TIM-2 Monoclonal antibody specifically detects TIM-2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
TIM-2
RMT2-26
PE
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2b, κ
RUO
Primary
Mouse