Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 (CD366) Mouse anti-Human, BB515, Clone: 7D3, BD Horizon™

Mouse Monoclonal Antibody

$150.00 - $345.00

Specifications

Antigen TIM-3 (CD366)
Isotype Mouse IgG1, κ
Immunogen Human TIM-3
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Conjugate BB515
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565569
View Documents
BD Biosciences
565569
25 Tests Each for $150.00
Add to cart
 
BDB565568
View Documents
BD Biosciences
565568
100 Tests Each for $345.00
Add to cart
 
Description

Description

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

TIM-3 (CD366)
Human TIM-3
BB515
Cell Biology
Murine
Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Mouse IgG1, κ
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
7D3
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3
Vol. Per test: 5μL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit