Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 (CD366) Mouse anti-Human, PE-CF594, Clone: 7D3, BD Horizon™

Mouse Monoclonal Antibody

$149.80 - $359.00

Specifications

Antigen TIM-3 (CD366)
Isotype Mouse IgG1 κ
Immunogen Human TIM-3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate PE-CF594
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565561
View Documents
BD Biosciences
565561
25 Tests
Each for $149.80
Add to cart
 
BDB565560
View Documents
BD Biosciences
565560
100 Tests
Each for $359.00
Add to cart
 
Description

Description

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

TIM-3 (CD366)
Human TIM-3
PE-CF594
Cell Biology
Mouse
Purified
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3
Vol. Per test: 5μL
Mouse IgG1 κ
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
7D3
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit