Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 (CD366) Mouse anti-Human, PerCP-Cy5.5, Clone: 7D3, BD Pharmingen™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 567123

 View more versions of this product

Catalog No. BDB567123


Add to Cart
This item is not returnable. View return policy

Description

Description

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIM-3 (CD366)
Monoclonal
PerCP-Cyanine5.5
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3
Human TIM-3
RUO
Human
Liquid
Flow Cytometry
7D3
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with PerCP-Cy5.5 under optimum conditions, and unconjugated antibody and free PerCP-Cy5.5 were removed. Storage of PerCP-Cy5.5 conjugates in unoptimized diluent is not recommended and may result in loss of signal intensity.
IgG1 κ
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit