Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, PerCP-Cy5.5, Clone: 7D3, BD Pharmingen™
Mouse Monoclonal Antibody
$160.00 - $380.00
Specifications
Antigen | TIM-3 (CD366) |
---|---|
Clone | 7D3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-Cyanine5.5 |
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow CytometrySpecifications
TIM-3 (CD366) | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
Mouse | |
Human | |
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with PerCP-Cy5.5 under optimum conditions, and unconjugated antibody and free PerCP-Cy5.5 were removed. Storage of PerCP-Cy5.5 conjugates in unoptimized diluent is not recommended and may result in loss of signal intensity. |
7D3 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human TIM-3 | |
Primary |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title