Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 Mouse anti-Human, Alexa Fluor™ 594, Clone: F38-2E2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227221AF594

Catalog No. NP227221A94


Add to cart

Description

Description

TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TIM-3
F38-2E2
Flow Cytometry, Flow (Cell Surface)
Conjugated, Purified
HAVCR2
This antibody was raised against recombinant human TIM-3. [UniprotNo. Q8TDQ0]
Protein G purified
Immunology, Innate Immunity
Primary
84868.0
Flow Cytometry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit