Learn More
TIM-3 Rabbit anti-Human, Alexa Fluor 350, Clone: 2321C, R&D Systems
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB23652U100UG
Catalog No. FB23652U100
Add to cart
This item is not returnable. View return policy
Description
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TIM-3
2321C
Flow Cytometry 0.25-1 μg/106 cells
Q8TDQ0
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession No. Q8TDQ0
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human TIM-3 in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
84868.0
Human