Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TIM-3 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB23652V100UG
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry.Specifications
TIM-3 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession No. Q8TDQ0 | |
100 μg | |
84868.0 | |
Human | |
Supernatant |
Flow Cytometry | |
2321C | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q8TDQ0 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human TIM-3 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction