Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 Rabbit anti-Mouse, PerCP, Clone: 2C12, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311989PCP
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Mouse samples. It is validated for Flow CytometrySpecifications
TIM-3 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG κ |
Flow Cytometry | |
2C12 | |
Flow Cytometry | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
This antibody was raised by immunising rats with TIM-3:Ig fusion protein emulsified in complete Freund's adjuvant. | |
0.1 mL | |
Immunology, Innate Immunity | |
84868 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title