Learn More
AdipoGenSupplier Diversity Partner TIM-4 (HU):FC(HU)(REC.)(100UG) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner CHIHF210T4C100
Tim-4 (human):Fc (human) (rec.), AdipoGen Life Sciences. Protein. The ed of human Tim-4 (aa 25-315) is fused to the N-terminus of the Fc region of human IgG1. Measured by its ability to inhibit anti-CD3-induced proliferation of stimulated human T cells. Lyophilized from 0.2�m-filtered solution in PBS.
Catalog No. 50-114-7382