Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM17 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551144

Catalog No. PA551144


Add to cart

Description

Description

The antibody detects endogenous levels of total TIM17 protein.

Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
Specifications

Specifications

TIM17
Polyclonal
Unconjugated
TIMM17A
Liquid
TIMM17A
Full length fusion protein of human TIMM17A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q99595, Q9Z0V8
TIM17, TIM17A
Rabbit
IgG
100 μL
-20°C
Primary
10440, 21854
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit