Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM50 Mouse anti-Human, Clone: OTI1E9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527499

Catalog No. PIMA527499


Add to cart

Description

Description

This gene encodes a subunit of the TIM23 inner mitochondrial membrane translocase complex. The encoded protein functions as the receptor subunit that recognizes the mitochondrial targeting signal, or presequence, on protein cargo that is destined for the mitochondrial inner membrane and matrix. This protein may also play a role in maintaining the membrane permeability barrier, and knockdown of this gene in human cells results in the release of cytochrome c and apoptosis.
Specifications

Specifications

TIMM50
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q3ZCQ8
TIMM50; TIM50L; translocase of inner mitochondrial membrane 50 homolog (yeast); Tim50-like protein; homolog of yeast Tim50; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); MGC102733; TIM50
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
92609
Western Blot
OTI1E9
Unconjugated
TIMM50
Liquid
TIMM50
Human recombit protein fragment corresponding to amino acids 45-353 of TIMM50 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit