Learn More
3M™ MonitorMark™ Time Temperature Indicator, Sold by AquaPhoenix Scientific
SCI_ED
Solves the challenge of monitoring thermal exposure of temperature-sensitive products during transportation and storage
Supplier:  3M™ MonitorMark™ 9860H
 View more versions of this product
Catalog No. S26884
Add to cart
Description
Description
  • Inexpensive solution for monitoring product exposure to temperatures above specified limit over 48 hours
  • Self-adhesive backing for easy attachment to secondary packaging
  • Easily-interpreted visual results
  • Results indicate both exposure and relative time over which exposure occurred
  • Removable film activation strip starts monitoring process on demand for accuracy
Specifications
Specifications
3.74 in.
1 week
88°F
95 mm
Time temperature indicator
31°C