Learn More
TIMM10B Mouse anti-Human, Clone: 1A11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524457
Catalog No. PIMA524457
Add to cart
Description
Description
Peptide Sequence: LRNLRDFLLV YNRMTELCFQ RCVPSLHHRA LDAEEEACLH SCAGKLIHSN HRLMAAYVQL MPALVQRRIA DYEAASAVPG VAAEQPGVSP SGS
FXC1, or TIMM10B, belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.
Specifications
Specifications
TIMM10B
Monoclonal
Unconjugated
TIMM10B
Liquid
TIMM10B
FXC1 (NP_036324.1, 11 a.a. ∽ 103 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
1A11
PBS with no preservative; pH 7.4
Q9Y5J6
TIMM10B; FXC1; TIM9B; TIMM9B
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
26515