Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Timolol Maleate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
300 g
Packaging:
Amber Glass Bottle
26921-17-5
C17H28N4O7S
432.49
WLRMANUAADYWEA-NWASOUNVSA-N
(2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1
26921-17-5
C17H28N4O7S
432.49
WLRMANUAADYWEA-NWASOUNVSA-N
(2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1

T1134, 26921-17-5, C13H24N4O3S•C4H4O4

Spectrum™ Timolol Maleate, USP is a non-selective beta-adrenergic receptor antagonist used to treat glaucoma, heart attacks, migraines, and hypertension. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Quantity 100 g
Ignition Residue 0.001
pH 3.8 to 4.3
CAS Max % 1
Percent Purity 98 to 102%
Grade USP