Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-1 Rabbit anti-Human, Clone: TIMP1/1944R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30771920UG

 View more versions of this product

Catalog No. NB122912


Add to cart

Description

Description

TIMP-1 Monoclonal specifically detects TIMP-1 in Human samples. It is validated for ELISA.
Specifications

Specifications

TIMP-1
TIMP1/1944R
Unconjugated
10 mM PBS with 0.05% BSA
CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Recombinant human full-length TIMP-1 protein
Protein A purified
RUO
Store at 4°C.
Monoclonal
Human
ELISA
0.2 mg/ml
ELISA 2-4 ug/ml
Purified
Rabbit
IgG
20 μg
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix
Primary
7076
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit