Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-1 Rabbit anti-Human, Clone: TIMP1/1944R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen TIMP-1
Clone TIMP1/1944R
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122911
View Documents
Novus Biologicals
NBP30771910UG
100 μg
Each for $406.52
Add to cart
 
NB122912
View Documents
Novus Biologicals
NBP30771920UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human samples. It is validated for ELISA
Specifications

Specifications

TIMP-1
0.2 mg/ml
ELISA
Purified
RUO
Human
CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Recombinant human full-length TIMP-1 protein
Monoclonal
Protein A purified
TIMP1/1944R
ELISA 2-4 ug/ml
Unconjugated
Rabbit
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix
10 mM PBS with 0.05% BSA
7076
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit