Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-2 Goat anti-Human, Clone: 40019A, R&D Systems™

Goat Monoclonal Antibody

$151.76 - $358.31

Specifications

Antigen TIMP-2
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Format Purified
Clone 40019A
Applications Western Blot, Immunohistochemistry, Immunoassay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB97112SP
View Documents Promotion Details
R&D Systems
MAB97112SP
25 ug
Each for $151.76
Add to cart
 
MAB97112100
View Documents Promotion Details
R&D Systems
MAB97112100
100 ug
Each for $358.31
Add to cart
 
Description

Description

TIMP-2 Monoclonal specifically detects TIMP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications

Specifications

TIMP-2
Purified
Western Blot, Immunohistochemistry, Immunoassay
Goat
Protein A or G purified from cell culture supernatant
RUO
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
40019A
Unconjugated
IgG
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit