Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-2 Goat anti-Human, Mouse, Polyclonal, R&D Systems
Goat Polyclonal Antibody
Manufacturer: R&D Systems AF971SP
This item is not returnable.
View return policy
Description
TIMP-2 Polyclonal antibody AF971SP specifically detects TIMP-2 in Human, Mouse samples. It is validated for Immunohistochemistry, Neutralization, Western BlottingSpecifications
TIMP-2 | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 2.6 ug/mL | |
P16035 | |
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
Goat | |
IgG | |
25 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human and mouse TIMP-2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TIMP-4 is observed. | |
Human, Mouse |
Immunohistochemistry, Neutralization, Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
TIMP2 | |
Chinese hamster ovary cell line CHO-derived recombinant human TIMP-2 Cys27-Pro220 Accession # P16035 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
7077 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only