Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-2 Mouse anti-Bovine, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish, Clone: 3A4- Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254555200UG
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
TIMP-2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
Mouse | |
21 kDa | |
200 μg | |
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers | |
7077 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
3A4 | |
Unconjugated | |
PBS with No Preservative | |
TIMP2 | |
A synthetic peptide from the N-terminus of human TIMP2 protein (aa56-66) (DSGNDIYGNPI) | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only