Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-2 Mouse anti-Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish, PE-Cyanine7, Clone: 3A4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254555PECY7

Catalog No. N25455PECY7


Add to cart

Description

Description

TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Flow Cytometry.
Specifications

Specifications

TIMP-2
3A4
Flow Cytometry
Conjugated, Purified
TIMP2
A synthetic peptide from the N-terminus of human TIMP2 protein (aa56-66) (DSGNDIYGNPI)
Protein A or G purified
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers
Primary
7077.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Bovine, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit