Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-4 Mouse anti-Human, Clone: MM0037-9F3, Novus Biologicals™
Mouse Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | TIMP-4 |
---|---|
Clone | MM0037-9F3 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11061003
|
Novus Biologicals
NB11061003 |
0.1 mg |
Each for $493.00
|
|
NB017995
|
Novus Biologicals
NB110610030.025MG |
0.025 mg |
Each for $177.16
|
|
Description
TIMP-4 Monoclonal specifically detects TIMP-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIMP-4 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
7079 | |
IgG1 | |
Protein G purified |
MM0037-9F3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cellular Markers, Hypoxia | |
metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
TIMP4 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title