Learn More
Abnova™ TIMP1 (Human) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5704
Catalog No. 89-339-635
Add to cart
Description
Description
TIMP1 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human TIMP1 in cell culture supernates, serum, plasma (heparin, EDTA) and saliva.
Specifications
Specifications
Quantitative
TIMP1
1 Kit
RUO
Human
7076
Standard curve
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
CLGI, EPA, EPO, FLJ90373, HCI, TIMP