Learn More
TIMP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA185923
Catalog No. PIPA185923
Add to Cart
Description
Description
This antibody does not neutralize the protein activity. Rabbit anti Human TIMP-1 antibody recognizes human Tissue inhibitor of metalloproteinases 1, also known as TIMP-1, Metalloproteinase inhibitor 1, Erythroid-potentiating activity or Fibroblast collagenase inhibitor.
TIMP1 Polyclonal antibody specifically detects TIMP1 in Bovine, Equine, Ferret, Human, Ovine, Rabbit, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIMP1
Polyclonal
Unconjugated
TIMP1
CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1
Rabbit
purified
RUO
100009047, 100034220, 116510, 282092, 443331, 7076
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O02722, P01033, P20414, P20614, P30120, P50122
TIMP1
Human tissue inhibitor of metalloproteinase 1 (TIMP-1).
100 μg
Primary
Bovine, Equine, Ferret, Human, Ovine, Rabbit, Rat
Antibody
IgG
Product Suggestions
Product Suggestions