Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP1 Rabbit anti-Human, Unconjugated, Clone: TIMP1/1944R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00641

Catalog No. 89-339-656


Add to cart

Description

Description

TIMP1 Monoclonal antibody specifically detects TIMP1 in Human samples. It is validated for ELISA.
Specifications

Specifications

TIMP1
TIMP1/1944R
TIMP metallopeptidase inhibitor 1
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
ELISA
Unconjugated
TIMP1
CLGI, EPA, EPO, FLJ90373, HCI, TIMP
Recombinant protein corresponding to full-length human TIMP1
100 μg
Primary
7076
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit