Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP1 Recombinant Rabbit Monoclonal Antibody (009), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$221.00 - $430.00
Specifications
Antigen | TIMP1 |
---|---|
Clone | 009 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543146
![]() |
Thermo Scientific
MA543146 |
100 μL |
Each for $430.00
|
|
PIMA543478
![]() |
Thermo Scientific
MA543478 |
50 μL |
Each for $221.00
|
|
Description
Specificity: Rat TIMP-1
TIMP1 Recombinant Monoclonal antibody specifically detects TIMP1 in Rat samples. It is validated for ELISASpecifications
TIMP1 | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1 | |
TIMP1 | |
IgG | |
Protein A | |
Antibody |
009 | |
ELISA | |
Unconjugated | |
Rabbit | |
Rat | |
P30120 | |
116510 | |
Recombinant Rat TIMP-1 Protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
TIMP1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title